Learn More
Analytical Sales & Services Daisy Chain Connector for Flow-Through LED array to TCR or Straight-Thru TTD
Catalog No. 5000804484
Encompass
Daisy Chain Connector
\n
Connects Lumidox II  LED Array with Flow-Through base to a Temperature Controlled Reactor (TCR) or Straight-Thru TTD
\n
\n
    \n
  • High-end, chemically resistant Tygon PVC tubing
  • \n
  • Tubing is clear, so you can easily monitor flow
  • \n
  • Ideal length to prevent tube kinking, while maintaining sufficient flexibility
  • \n
  • Tubing terminated on both ends with precise hose-barbed inserts for superior grip
  • \n
  • Temperature range -20°C to 70°C (please contact us for temperature requirements outside this range)
  • \n
\n
\n \n\n 
Catalog No. 50-008-04484 Supplier Analytical Sales & Services Supplier No. 266704
Daisy Chain Connector
\n
Connects Lumidox II  LED Array with Flow-Through base to a Temperature Controlled Reactor (TCR) or Straight-Thru TTD
\n
\n
    \n
  • High-end, chemically resistant Tygon PVC tubing
  • \n
  • Tubing is clear, so you can easily monitor flow
  • \n
  • Ideal length to prevent tube kinking, while maintaining sufficient flexibility
  • \n
  • Tubing terminated on both ends with precise hose-barbed inserts for superior grip
  • \n
  • Temperature range -20°C to 70°C (please contact us for temperature requirements outside this range)
  • \n
\n
\n \n\n 