Learn More
Analytical Sales & Services Daisy Chain Connector for TCR or Straight-Thru TTD to TTD+ or Flow Reactor
Catalog No. 5000804308
Encompass
Daisy Chain Connector
\n
Connects TCR or Straight-Thru TTD to TTD+ or Flow Reactor
\n
\n
    \n
  • High-end, chemically resistant Tygon PVC tubing
  • \n
  • Tubing is clear, so you can easily monitor flow
  • \n
  • Ideal length to prevent tube kinking, while maintaining sufficient flexibility
  • \n
  • Tubing terminated on both ends with precise hose-barbed inserts for superior grip
  • \n
  • Temperature range -20°C to 70°C (please contact us for temperature requirements outside this range)
  • \n
\n
\n 
Catalog No. 50-008-04308 Supplier Analytical Sales & Services Supplier No. 266706
Daisy Chain Connector
\n
Connects TCR or Straight-Thru TTD to TTD+ or Flow Reactor
\n
\n
    \n
  • High-end, chemically resistant Tygon PVC tubing
  • \n
  • Tubing is clear, so you can easily monitor flow
  • \n
  • Ideal length to prevent tube kinking, while maintaining sufficient flexibility
  • \n
  • Tubing terminated on both ends with precise hose-barbed inserts for superior grip
  • \n
  • Temperature range -20°C to 70°C (please contact us for temperature requirements outside this range)
  • \n
\n
\n 