Learn More
Analytical Sales & Services Daisy Chain/Tubing Union
Catalog No. 5000806066
Encompass
Daisy Chain/Tubing Union
\n
Use to join:
\n
• Two 266701 Daisy Chains together, or
\n
• Two lengths of tubing that come with the TCubeEdge, or
\n
• 266701 Daisy Chain to the tubing that comes with the TCubeEdge
\n 
Catalog No. 50-008-06066 Supplier Analytical Sales & Services Supplier No. 266707
Daisy Chain/Tubing Union
\n
Use to join:
\n
• Two 266701 Daisy Chains together, or
\n
• Two lengths of tubing that come with the TCubeEdge, or
\n
• 266701 Daisy Chain to the tubing that comes with the TCubeEdge
\n 