Learn More
CPC Scientific H-Tyr-DAla-Phe-Asp-Val-Val-Gly-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OPIO001A
Encompass
Selective delta-opoid receptor agonist isolated from the skin of Phyllomedusa Bicolor.SEQUENCE: H-Tyr-DAla-Phe-Asp-Val-Val-Gly-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Y-DAla-FDVVG-NH2MOLECULAR FORMULA: C37H52N8O10MOLECULAR WEIGHT: 768.87STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [122752-15-2]SYNONYMS: Deltorphin I, Deltorphin CRESEARCH AREA: PainREFERENCES:SKU(s): OPIO-001A
Catalog No. 50-210-9972