Learn More
CPC Scientific H-Tyr-DAla-Phe-Glu-Val-Val-Gly-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OPIO002A
Encompass
Selective delta-opoid receptor agonist isolated from the skin of Phyllomedusa Bicolor.SEQUENCE: H-Tyr-DAla-Phe-Glu-Val-Val-Gly-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Y-DAla-FEVVG-NH2MOLECULAR FORMULA: C38H54N8O10MOLECULAR WEIGHT: 782.90STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [122752-16-3]SYNONYMS: Deltorphin B, Deltorphin IIRESEARCH AREA: PainREFERENCES:SKU(s): OPIO-002A
Catalog No. 50-210-9973