Learn More
TARGETMOL CHEMICALS INC DALBAVANCIN 5MG
SDP
Catalog No. 502577219
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dalbavancin (BI-397) a novel second-generation lipoglycopeptide antibiotic possesses in vitro activity against a variety of Gram-positive pathogens including methicillin-resistant Staphylococcus aureus (MRSA) and methicillin-resistant Staphylococcus epidermidis (MRSE)Dalbavancin inhibits Staphylococcus aureus and Bacillus anthracis with MIC90s of 0.06 ug/mL and 0.25 ug/mL respectivelypurity: 98%
Catalog No. 50-257-7219 Supplier TARGETMOL CHEMICALS INC Supplier No. T213585MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dalbavancin (BI-397) a novel second-generation lipoglycopeptide antibiotic possesses in vitro activity against a variety of Gram-positive pathogens including methicillin-resistant Staphylococcus aureus (MRSA) and methicillin-resistant Staphylococcus epidermidis (MRSE)Dalbavancin inhibits Staphylococcus aureus and Bacillus anthracis with MIC90s of 0.06 ug/mL and 0.25 ug/mL respectivelypurity: 98%