Learn More
Selleck Chemical LLC Dalbavancin S4848-2mg
SDP
Catalog No. 503108214
Encompass_Preferred
Dalbavancin is a lipoglycopeptide antibiotic that has bactericidal activity against Gram-positive bacteria including various staphylococci
Catalog No. 50-310-8214 Supplier Selleck Chemical LLC Supplier No. S48482MG
Dalbavancin is a lipoglycopeptide antibiotic that has bactericidal activity against Gram-positive bacteria including various staphylococci