Learn More
Medchemexpress LLC HY-17586 5mg Medchemexpress, Dalbavancin (hydrochloride) CAS:2227366-51-8 Purity:>98%
SDP
Catalog No. 502081993
Encompass_Preferred
Medchemexpress, HY-17586 5mg Dalbavancin (hydrochloride) CAS:2227366-51-8 Dalbavancin hydrochloride (MDL-63397 hydrochloride) is a semisynthetic lipoglycopeptide antibiotic with potent bactericidal activity against Gram-positive bacteria. Dalbavancin hydrochloride inhibits Staphylococcus aureus and Bacillus anthracis with MIC90s of 0.06 μg/mL and 0.25 μg/mL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1993 Supplier Medchemexpress LLC Supplier No. HY175865MG
Medchemexpress, HY-17586 5mg Dalbavancin (hydrochloride) CAS:2227366-51-8 Dalbavancin hydrochloride (MDL-63397 hydrochloride) is a semisynthetic lipoglycopeptide antibiotic with potent bactericidal activity against Gram-positive bacteria. Dalbavancin hydrochloride inhibits Staphylococcus aureus and Bacillus anthracis with MIC90s of 0.06 μg/mL and 0.25 μg/mL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.