Learn More
Medchemexpress LLC Dalbavancin hydrochloride | 2227366-51-8 | MDL-63397 | 99.7% | 1853.15 | C88H101Cl3N10O28 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1758610MG
Encompass_Preferred
Dalbavancin hydrochloride is a semisynthetic lipoglycopeptide antibiotic provided as the hydrochloride salt for research use. It is active against gram-positive bacteria, including MRSA, and is supplied as a high-purity solid for laboratory assays.
  • High purity (99.7%).
  • White to light-yellow solid appearance.
  • Molecular weight 1853.15.
  • Active against gram-positive bacteria, including MRSA.
  • Suitable for in vitro antibacterial and pharmacological studies.
Catalog No. 50-002-97955