Learn More
Cayman Chemical DalcetrapIb 10mg
SDP
Catalog No. 502893809
Encompass_Preferred
An inhibitor of CETP; inhibits plasma CETP with an IC50 value of 9 µM; in rabbits given an atherogenic diet, increases HDL, decreases VLDL, and attenuates induced atherosclerosis; in humans, inhibits CETP activity and increases HDL levels but does not reduce the risk of recurrent cardiovascular events
Catalog No. 50-289-3809 Supplier Cayman Chemical Supplier No. 8945010 MG
An inhibitor of CETP; inhibits plasma CETP with an IC50 value of 9 µM; in rabbits given an atherogenic diet, increases HDL, decreases VLDL, and attenuates induced atherosclerosis; in humans, inhibits CETP activity and increases HDL levels but does not reduce the risk of recurrent cardiovascular events