Learn More
Medchemexpress LLC Dalcetrapib | 211513-37-0 | MFCD06407886 | 99.2% | 389.59 | 50 MG
SDP
Catalog No. 502081665
Encompass_Preferred
Dalcetrapib is an orally active cholesteryl ester transfer protein (CETP) inhibitor. It has shown activity against recombinant human CETP and human plasma CETP, playing a role in cholesterol metabolism research.
  • Inhibits cholesteryl ester transfer protein activity
  • Increases pre-β-HDL formation
  • Increases plasma HDL cholesterol
  • Promotes fecal elimination of neutral sterols and bile acids
Catalog No. 50-208-1665 Supplier Medchemexpress LLC Supplier No. HY1495050MG
Dalcetrapib is an orally active cholesteryl ester transfer protein (CETP) inhibitor. It has shown activity against recombinant human CETP and human plasma CETP, playing a role in cholesterol metabolism research.
  • Inhibits cholesteryl ester transfer protein activity
  • Increases pre-β-HDL formation
  • Increases plasma HDL cholesterol
  • Promotes fecal elimination of neutral sterols and bile acids