Learn More
Selleck Chemical LLC Dalcetrapib (JTT-705) S2772-5mg
SDP
Catalog No. 503112444
Encompass_Preferred
Dalcetrapib (JTT-705) is a rhCETP inhibitor with IC50 of 0 2 M that increases the plasma HDL cholesterol Phase 3
Catalog No. 50-311-2444 Supplier Selleck Chemical LLC Supplier No. S27725MG
Dalcetrapib (JTT-705) is a rhCETP inhibitor with IC50 of 0 2 M that increases the plasma HDL cholesterol Phase 3