Learn More
TARGETMOL CHEMICALS INC DALCOTIDINE 5MG
SDP
Catalog No. 502580375
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dalcotidine (KU 1257) is a novel histamine H2 receptor antagonist with histamine H9 receptor antagonistic activity and antisecretory effects.The Ki value for binding to guinea pig cerebral cortex was 0.040 The KB value for antagonism of histamine-induced positive chronotropic responses in the right atrium of isolate guinea pigs was 0.041.Dalcotidine improves the quality of ulcer healing and may contribute to a reduction in ulcer recurrence and relapse rates. purity: 98%
Catalog No. 50-258-0375 Supplier TARGETMOL CHEMICALS INC Supplier No. T311955MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dalcotidine (KU 1257) is a novel histamine H2 receptor antagonist with histamine H9 receptor antagonistic activity and antisecretory effects.The Ki value for binding to guinea pig cerebral cortex was 0.040 The KB value for antagonism of histamine-induced positive chronotropic responses in the right atrium of isolate guinea pigs was 0.041.Dalcotidine improves the quality of ulcer healing and may contribute to a reduction in ulcer recurrence and relapse rates. purity: 98%