Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dalda (acetate) | 99.8% | 671.79 | 5 MG
SDP

Catalog No. 5000399322
Encompass_Preferred

DALDA acetate is a potent and highly selective μ-opioid receptor agonist with a Ki of 1.69 nM. It exhibits antinociceptive and respiratory effects. This compound carries a net positive charge (3+) at physiological pH, making it hydrophilic and more polar than morphine.

  • Potent and highly selective μ-opioid receptor agonist
  • Exhibits antinociceptive and respiratory effects
  • Hydrophilic and more polar than morphine
  • Appearance: Solid, white to off-white
  • Purity: 99.8%

Catalog No. 50-003-99322 Supplier Medchemexpress LLC Supplier No. HYP3870A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DALDA acetate is a potent and highly selective μ-opioid receptor agonist with a Ki of 1.69 nM. It exhibits antinociceptive and respiratory effects. This compound carries a net positive charge (3+) at physiological pH, making it hydrophilic and more polar than morphine.

  • Potent and highly selective μ-opioid receptor agonist
  • Exhibits antinociceptive and respiratory effects
  • Hydrophilic and more polar than morphine
  • Appearance: Solid, white to off-white
  • Purity: 99.8%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.