Learn More
Medchemexpress LLC Dalidnetug 10mg | 10mg
SDP
Catalog No. 5000323228
Encompass_Preferred
Dalidnetug is a humanized monoclonal antibody targeting human amyloid-beta precursor protein (APP) Dalidnetug specifically binds to APP to reduce the production of amyloid-beta (A ) thus exerting the activity of clearing amyloid-beta Dalidnetug is promising for research of Alzheimer s disease[1]
Catalog No. 50-003-23228 Supplier Medchemexpress LLC Supplier No. HYP99113010MG
Dalidnetug is a humanized monoclonal antibody targeting human amyloid-beta precursor protein (APP) Dalidnetug specifically binds to APP to reduce the production of amyloid-beta (A ) thus exerting the activity of clearing amyloid-beta Dalidnetug is promising for research of Alzheimer s disease[1]