Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dalpiciclib 10mg | 1637781-04-4 | 10 MG
SDP

Catalog No. 5000228092
Encompass_Preferred

Dalpiciclib is an orally active, selective cyclin-dependent kinase 4 and 6 (CDK4/6) inhibitor supplied for research use in a 10 mg vial. It is intended for preclinical and in vitro studies of cell-cycle regulation and anti-tumor activity.

  • Selective CDK4/6 inhibitor used in preclinical research.
  • Cas 1637781-04-4; molecular formula C25H30N6O2.
  • Molecular weight 446.54 g/mol.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months).
  • Soluble in DMSO (50 mg/mL); manufacturer lists in vivo vehicle formulations for dosing.

Catalog No. 50-002-28092 Supplier Medchemexpress LLC Supplier No. HY11433810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dalpiciclib is an orally active, selective cyclin-dependent kinase 4 and 6 (CDK4/6) inhibitor supplied for research use in a 10 mg vial. It is intended for preclinical and in vitro studies of cell-cycle regulation and anti-tumor activity.

  • Selective CDK4/6 inhibitor used in preclinical research.
  • Cas 1637781-04-4; molecular formula C25H30N6O2.
  • Molecular weight 446.54 g/mol.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months).
  • Soluble in DMSO (50 mg/mL); manufacturer lists in vivo vehicle formulations for dosing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.