Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dalpiciclib 25mg | 1637781-04-4 | 25 MG
SDP

Catalog No. 5000226570
Encompass_Preferred

Dalpiciclib is a selective, orally active inhibitor of cyclin-dependent kinases 4 and 6 used in preclinical research to study cell-cycle regulation and anticancer activity. It is supplied as an off-white to light yellow solid with recommended storage and solubility handling for laboratory use.

  • High purity for research applications.
  • Molecular weight 446.54 g/mol and formula C25H30N6O2.
  • Soluble in DMSO with recommended handling and warming.
  • Stable when stored as powder at -20°C for long-term storage.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-26570 Supplier Medchemexpress LLC Supplier No. HY11433825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dalpiciclib is a selective, orally active inhibitor of cyclin-dependent kinases 4 and 6 used in preclinical research to study cell-cycle regulation and anticancer activity. It is supplied as an off-white to light yellow solid with recommended storage and solubility handling for laboratory use.

  • High purity for research applications.
  • Molecular weight 446.54 g/mol and formula C25H30N6O2.
  • Soluble in DMSO with recommended handling and warming.
  • Stable when stored as powder at -20°C for long-term storage.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.