Learn More
Medchemexpress LLC Dalpiciclib 50mg | 1637781-04-4 | 50 MG
SDP
Catalog No. 5000229925
Encompass_Preferred
Dalpiciclib is an orally active, selective inhibitor of cyclin-dependent kinases CDK4 and CDK6 intended for preclinical research use. It is supplied as a powder and is characterized by CAS 1637781-04-4, molecular formula C25H30N6O2, and molecular weight 446.54 g/mol. For research use only; not for human or clinical use.
  • Selective CDK4/6 inhibition with low-nanomolar potency
  • High chemical purity suitable for analytical and biological studies
  • Supplied as a powder for flexible formulation and dosing
  • Stable under recommended storage conditions for long-term use
  • Characterized by validated analytical data and CAS identification
Catalog No. 50-002-29925 Supplier Medchemexpress LLC Supplier No. HY11433850MG
Dalpiciclib is an orally active, selective inhibitor of cyclin-dependent kinases CDK4 and CDK6 intended for preclinical research use. It is supplied as a powder and is characterized by CAS 1637781-04-4, molecular formula C25H30N6O2, and molecular weight 446.54 g/mol. For research use only; not for human or clinical use.
  • Selective CDK4/6 inhibition with low-nanomolar potency
  • High chemical purity suitable for analytical and biological studies
  • Supplied as a powder for flexible formulation and dosing
  • Stable under recommended storage conditions for long-term use
  • Characterized by validated analytical data and CAS identification