Learn More
Medchemexpress LLC Dalpiciclib | 5MG
SDP
Catalog No. 5000228676
Encompass_Preferred
Dalpiciclib (SHR-6390) is an orally active and highly selective inhibitor of CDK4 and 6 with IC50 values of 12 4 nM and 9 9 nM respectively[1][2] Dalpiciclib shows antitumor activity against breast cancer and esophageal squamous cell carcinoma[1][2][3][4]
Catalog No. 50-002-28676 Supplier Medchemexpress LLC Supplier No. HY1143385MG
Dalpiciclib (SHR-6390) is an orally active and highly selective inhibitor of CDK4 and 6 with IC50 values of 12 4 nM and 9 9 nM respectively[1][2] Dalpiciclib shows antitumor activity against breast cancer and esophageal squamous cell carcinoma[1][2][3][4]