Learn More
TARGETMOL CHEMICALS INC DALTROBAN 5MG
SDP
Catalog No. 502581256
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Daltroban (SKF 96148) is a specific thromboxane A2 (TXA2) receptor antagonist. Daltroban shows protective effect in reperfusion injury. Daltroban increase intracellular calcium in vascular smooth muscle cells. purity: 99%
Catalog No. 50-258-1256 Supplier TARGETMOL CHEMICALS INC Supplier No. T150495MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Daltroban (SKF 96148) is a specific thromboxane A2 (TXA2) receptor antagonist. Daltroban shows protective effect in reperfusion injury. Daltroban increase intracellular calcium in vascular smooth muscle cells. purity: 99%