Learn More
TARGETMOL CHEMICALS INC DALVASTATIN 5MG
SDP
Catalog No. 502580760
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Dalvastatin (RG-12561) is an orally available inhibitor of HMG-CoA reductase and cholesterol-lowering synthesis.Dalvastatin competitively inhibits rat hepatic HMG-CoA reductase with an IC50 value of 3.4 nmol / l. Dalvastatin inhibits cholesterol biosynthesis in liver sections in rat experiments with an ED50 value of 0.9. purity: 98%
Catalog No. 50-258-0760 Supplier TARGETMOL CHEMICALS INC Supplier No. T252855MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Dalvastatin (RG-12561) is an orally available inhibitor of HMG-CoA reductase and cholesterol-lowering synthesis.Dalvastatin competitively inhibits rat hepatic HMG-CoA reductase with an IC50 value of 3.4 nmol / l. Dalvastatin inhibits cholesterol biosynthesis in liver sections in rat experiments with an ED50 value of 0.9. purity: 98%