Learn More
New England Biolabs, Inc. dam-/dcm- Competent E. coli – 20 x 0.05 ml/tube
SDP
Catalog No. 50995106
Encompass
Methyltransferase deficient chemically competent E. coli cells suitable for growth of plasmids free of Dam and Dcm methylation. Note that dam- strains are not recommended as a host for primary cloning/ligation. The dam mutation can result in an increased mutation rate in the cell and a reduction in the transformation efficiency. DNA should be maintained in a dam+ strain unless there is a specific need for DNA free of Dam or Dcm methylation.
Catalog No. 50-995-106 Supplier New England Biolabs, Inc. Supplier No. C2925H
Methyltransferase deficient chemically competent E. coli cells suitable for growth of plasmids free of Dam and Dcm methylation. Note that dam- strains are not recommended as a host for primary cloning/ligation. The dam mutation can result in an increased mutation rate in the cell and a reduction in the transformation efficiency. DNA should be maintained in a dam+ strain unless there is a specific need for DNA free of Dam or Dcm methylation.