Learn More
New England Biolabs, Inc. dam Methyltransferase – 2500 units
SDP
Catalog No. 50811641
Encompass
dam Methyltransferase modifies the adenine residue (N6) of the sequence GATC.
Catalog No. 50-811-641 Supplier New England Biolabs, Inc. Supplier No. M0222L
dam Methyltransferase modifies the adenine residue (N6) of the sequence GATC.