Learn More
AdipoGen 7-Dimethylamino-4-methylcoumarin (5 g)
SDP
Supplier:  AdipoGen CDXD0711G005
Encompass_Preferred
Chemical. CAS: 87-01-4. Formula: C12H13NO2. MW: 203.24. Synthetic. Fluorescent probe. Spectral data (EtOH): Em/Ex= 366/447 nm). Intermediate/Reactant for organic synthesis of dyes. Used as brightener in textile industry and as laser dye.
Catalog No. 50-205-4397