Learn More
Medchemexpress LLC HY-P0210 10mg , DAMGO CAS:78123-71-4 Purity:>98%
SDP
Catalog No. 502261204
Encompass_Preferred
HY-P0210 10mg DAMGO CAS: 78123-71-4 DAMGO is a μ-opioid receptor (μ-OPR ) selective agonist with a Kd of 3.46 nM for native μ-OPR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1204 Supplier Medchemexpress LLC Supplier No. HYP021010MG
HY-P0210 10mg DAMGO CAS: 78123-71-4 DAMGO is a μ-opioid receptor (μ-OPR ) selective agonist with a Kd of 3.46 nM for native μ-OPR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.