Learn More
Medchemexpress LLC HY-P0210B 1mg Medchemexpress, DAMGO (TFA) CAS:950492-85-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0210B1MG
Encompass_Preferred
Medchemexpress, HY-P0210B 1mg DAMGO (TFA) CAS:950492-85-0 DAMGO TFA is a μ-opioid receptor ( μ-OPR ) selective agonist with a K d of 3.46 nM for native μ-OPR [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4053