Learn More
Cayman Chemical DAMGOtrIfluoroacetAT salt 10mg
SDP
Catalog No. 502987502
Encompass_Preferred
A selective peptide agonist of the µ-opioid receptor (Kis = 1.18, 1,430, and 213 nM for human µ-, δ-, and κ-opioid receptors, respectively)
Catalog No. 50-298-7502 Supplier Cayman Chemical Supplier No. 2155310 MG
A selective peptide agonist of the µ-opioid receptor (Kis = 1.18, 1,430, and 213 nM for human µ-, δ-, and κ-opioid receptors, respectively)