Learn More
Cayman Chemical DAmIno ACD OxIdase InhIbIto 5g
SDP
Catalog No. 502994454
Encompass_Preferred
An inhibitor of FGFR4 (IC50 1 3 nM) selective for FGFR4 over FGFR1 and a panel of 36 kinases (IC50s 10 UM for all) inhibits proliferation of Hep3B and Huh7 hepatic cancer cells with IC50 values of 1 1 and 2 5 nM respectively reduces tumor growth by 62 7 and 70 8 in a Huh7 mouse xenograft model at 30 and 100 mg/kg per day respectively
Catalog No. 50-299-4454 Supplier Cayman Chemical Supplier No. 238945 G
An inhibitor of FGFR4 (IC50 1 3 nM) selective for FGFR4 over FGFR1 and a panel of 36 kinases (IC50s 10 UM for all) inhibits proliferation of Hep3B and Huh7 hepatic cancer cells with IC50 values of 1 1 and 2 5 nM respectively reduces tumor growth by 62 7 and 70 8 in a Huh7 mouse xenograft model at 30 and 100 mg/kg per day respectively