Learn More
Medchemexpress LLC Daminozide | 1596-84-5 | 99.69% | 160.17 | 50 MG
SDP
Catalog No. 5000351600
Encompass_Preferred
Daminozide (Standard) is the analytical standard of Daminozide, designed for research and analytical applications. This compound acts as a plant growth regulator and is a selective inhibitor of human KDM2/7 histone demethylases. It demonstrates significant selectivity for the KDM2/7 subfamily, making it a valuable tool for specific biochemical investigations.
  • Analytical standard for research and analytical applications.
  • Functions as a plant growth regulator.
  • Selectively inhibits human KDM2/7 histone demethylases.
  • Exhibits over 100-fold selectivity for the KDM2/7 subfamily.
  • Used as a reference standard in various assays.
  • Applicable in qualitative, quantitative, and methodological research experiments.
  • Typically appears as a white to off-white solid.
Catalog No. 50-003-51600 Supplier Medchemexpress LLC Supplier No. HY13643R50MG
Daminozide (Standard) is the analytical standard of Daminozide, designed for research and analytical applications. This compound acts as a plant growth regulator and is a selective inhibitor of human KDM2/7 histone demethylases. It demonstrates significant selectivity for the KDM2/7 subfamily, making it a valuable tool for specific biochemical investigations.
  • Analytical standard for research and analytical applications.
  • Functions as a plant growth regulator.
  • Selectively inhibits human KDM2/7 histone demethylases.
  • Exhibits over 100-fold selectivity for the KDM2/7 subfamily.
  • Used as a reference standard in various assays.
  • Applicable in qualitative, quantitative, and methodological research experiments.
  • Typically appears as a white to off-white solid.