Learn More
Medchemexpress LLC Daminozide | 1596-84-5 | 99.8% | 160.17 | 5 G
SDP
Catalog No. 5000350278
Encompass_Preferred
Daminozide is a plant growth regulator and a selective inhibitor of human KDM2/7 histone demethylases. It has IC50s of 0.55, 1.5, and 2.1 μM for PHF8, KDM2A, and KIAA1718, respectively. Daminozide demonstrates over 100-fold selectivity for the KDM2/7 subfamily compared to other demethylase subfamily members tested. It is intended for research use only.
  • Plant growth regulator
  • Selective inhibitor of human KDM2/7 histone demethylases
  • High selectivity for KDM2/7 subfamily (>100-fold)
Catalog No. 50-003-50278 Supplier Medchemexpress LLC Supplier No. HY136435G
Daminozide is a plant growth regulator and a selective inhibitor of human KDM2/7 histone demethylases. It has IC50s of 0.55, 1.5, and 2.1 μM for PHF8, KDM2A, and KIAA1718, respectively. Daminozide demonstrates over 100-fold selectivity for the KDM2/7 subfamily compared to other demethylase subfamily members tested. It is intended for research use only.
  • Plant growth regulator
  • Selective inhibitor of human KDM2/7 histone demethylases
  • High selectivity for KDM2/7 subfamily (>100-fold)