Learn More
Medchemexpress LLC Daminozide | 1596-84-5 | MFCD00002787 | 99.8% | 160.17 | 1 G
SDP
Catalog No. 5000352376
Encompass_Preferred
Daminozide is a plant growth regulator that acts as a selective inhibitor of human KDM2/7 histone demethylases. It demonstrates significant selectivity for the KDM2/7 subfamily over other demethylase subfamily members, making it a valuable tool in epigenetics research.
  • Selective inhibitor of human KDM2/7 histone demethylases
  • IC50s: 0.55 μM (PHF8), 1.5 μM (KDM2A), 2.1 μM (KIAA1718)
  • Exhibits >100-fold selectivity for KDM2/7 subfamily
Catalog No. 50-003-52376 Supplier Medchemexpress LLC Supplier No. HY136431G
Daminozide is a plant growth regulator that acts as a selective inhibitor of human KDM2/7 histone demethylases. It demonstrates significant selectivity for the KDM2/7 subfamily over other demethylase subfamily members, making it a valuable tool in epigenetics research.
  • Selective inhibitor of human KDM2/7 histone demethylases
  • IC50s: 0.55 μM (PHF8), 1.5 μM (KDM2A), 2.1 μM (KIAA1718)
  • Exhibits >100-fold selectivity for KDM2/7 subfamily