Learn More
Selleck Chemical LLC Daminozide S4800-500mg
SDP
Catalog No. 503109201
Encompass_Preferred
Daminozide (Aminozide DMASA DIMG B 995 Alar Kylar B-NINE SADH) a plant growth regulator selectively inhibits the KDM2/7 JmjC subfamily with IC50 of 1 5 0 7 M for KDM2A It is at least 100-fold selective as an inhibitor of the KDM2/7 subfamily over the other demethylase subfamily members tested with IC50s of 2 M or less against KDM2A PHF8 and KIAA1718 and IC50s of 127 M for KDM3A or greater (mM range) against other demethylases
Catalog No. 50-310-9201 Supplier Selleck Chemical LLC Supplier No. S4800500MG
Daminozide (Aminozide DMASA DIMG B 995 Alar Kylar B-NINE SADH) a plant growth regulator selectively inhibits the KDM2/7 JmjC subfamily with IC50 of 1 5 0 7 M for KDM2A It is at least 100-fold selective as an inhibitor of the KDM2/7 subfamily over the other demethylase subfamily members tested with IC50s of 2 M or less against KDM2A PHF8 and KIAA1718 and IC50s of 127 M for KDM3A or greater (mM range) against other demethylases