Learn More
Medchemexpress LLC HY-13643 500mg Medchemexpress, Daminozide CAS:1596-84-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13643500MG
Encompass_Preferred
Medchemexpress, HY-13643 500mg Daminozide CAS:1596-84-5 Daminozide, a plant growth regulator, is a selective inhibitor of the human KDM2/7 histone demethylases, with IC50s of 0.55, 1.5 and 2.1 μM for PHF8, KDM2A, and KIAA1718, respectively. Daminozide has >100-fold selectivity for KDM2/7 subfamily versus other demethylase subfamily members tested. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1441