Learn More
Cayman Chemical Daminozide-d4, Purity: ≥99% deuterated forms (d1-d4), 10 mg
SDP
Catalog No. 502915732
Encompass_Preferred
An internal standard for the quantification of daminozide by GC- or LC-MS
Catalog No. 50-291-5732 Supplier Cayman Chemical Supplier No. 3926410 MG
An internal standard for the quantification of daminozide by GC- or LC-MS