Learn More
Cayman Chemical Daminozide-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502916584
Encompass_Preferred
An internal standard for the quantification of daminozide by GC- or LC-MS
Catalog No. 50-291-6584 Supplier Cayman Chemical Supplier No. 392641 MG
An internal standard for the quantification of daminozide by GC- or LC-MS