Learn More
Medchemexpress LLC Daminozide (Standard) | 1596-84-5 | 99.79% | 160.17 | 250 MG
SDP
Catalog No. 5000348245
Encompass_Preferred
Daminozide (Standard) serves as an analytical standard for Daminozide, designed for research and analytical applications. This plant growth regulator selectively inhibits human KDM2/7 histone demethylases, showing over 100-fold selectivity compared to other demethylase subfamily members. It is widely utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard
  • Plant growth regulator
  • Selective inhibitor of human KDM2/7 histone demethylases
  • Utilized in HPLC, GC, and MS research
  • White to off-white solid
Catalog No. 50-003-48245 Supplier Medchemexpress LLC Supplier No. 250MGHY13643R
Daminozide (Standard) serves as an analytical standard for Daminozide, designed for research and analytical applications. This plant growth regulator selectively inhibits human KDM2/7 histone demethylases, showing over 100-fold selectivity compared to other demethylase subfamily members. It is widely utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard
  • Plant growth regulator
  • Selective inhibitor of human KDM2/7 histone demethylases
  • Utilized in HPLC, GC, and MS research
  • White to off-white solid