Learn More
Medchemexpress LLC Daminozide (Standard) | 1596-84-5 | 99.8% | 160.17 | 25 MG
SDP
Catalog No. 5000347893
Encompass_Preferred
Daminozide (Standard) serves as the analytical standard for Daminozide, a plant growth regulator and selective inhibitor of human KDM2/7 histone demethylases. It is intended for research and analytical applications, offering high selectivity for the KDM2/7 subfamily.
  • Used as a reference standard for assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments
  • Suitable for use in HPLC, GC, and MS applications
Catalog No. 50-003-47893 Supplier Medchemexpress LLC Supplier No. 25MGHY13643R
Daminozide (Standard) serves as the analytical standard for Daminozide, a plant growth regulator and selective inhibitor of human KDM2/7 histone demethylases. It is intended for research and analytical applications, offering high selectivity for the KDM2/7 subfamily.
  • Used as a reference standard for assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments
  • Suitable for use in HPLC, GC, and MS applications