Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daminozide 98.0+%, TCI America™
SDP

Catalog No. D40155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40155G 5 g
D401525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40155G Supplier TCI America Supplier No. D40155G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1596-84-5
Molecular Formula C6H12N2O3
Molecular Weight (g/mol) 160.17
MDL Number MFCD00002787
InChI Key NOQGZXFMHARMLW-UHFFFAOYSA-N
Synonym daminozide, kylar, alar, dmasa, aminozide, n-dimethylamino succinamic acid, sadh, b-nine, dimas, succinic acid 2,2-dimethylhydrazide
PubChem CID 15331
ChEBI CHEBI:4312
IUPAC Name 3-(N',N'-dimethylhydrazinecarbonyl)propanoic acid
SMILES CN(C)NC(=O)CCC(O)=O

Specifications

Melting Point 155°C
Color White
pH 3.8 (5% H2O)
Quantity 5 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Daminozide
RTECSNumber WM9625000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.