Learn More
American Radiolabeled Chemicals Inc DAMP 2 8-3H 1LC
SDP
Catalog No. 50901204
Encompass_Preferred
Deoxyadenosine 5'-2,8-3H monophosphate tetra(triethylammonium) salt 1mCi in ethanol:water (1:1), 1 mCi/ml, S.A. 10-25 Ci/mmol, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-204 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 17801MC
Deoxyadenosine 5'-2,8-3H monophosphate tetra(triethylammonium) salt 1mCi in ethanol:water (1:1), 1 mCi/ml, S.A. 10-25 Ci/mmol, Shipped in Dry Ice, Exclusive