Learn More
American Radiolabeled Chemicals Inc DAMP 2 8-3H 250UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 1780250UC
Encompass_Preferred
Deoxyadenosine 5'-2,8-3H monophosphate tetra(triethylammonium) salt 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 10-25 Ci/mmol, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-205
This product is regulated by the federal government, and may not be ordered online.