Learn More
Medchemexpress LLC Damulin A | 1202868-74-3 | 96.9% | 783.00 | 1 MG
SDP
Catalog No. 5000391170
Encompass_Preferred
Damulin A is a saponin found in G. pentaphyllum that exhibits anti-cancer activities. It shows stronger cytotoxicity against A549 cells with an IC50 value of 59.2 μM.
  • A saponin found in G. pentaphyllum
  • Possesses anti-cancer activities
  • Demonstrates cytotoxicity against A549 cells
Catalog No. 50-003-91170 Supplier Medchemexpress LLC Supplier No. HY169411MG
Damulin A is a saponin found in G. pentaphyllum that exhibits anti-cancer activities. It shows stronger cytotoxicity against A549 cells with an IC50 value of 59.2 μM.
  • A saponin found in G. pentaphyllum
  • Possesses anti-cancer activities
  • Demonstrates cytotoxicity against A549 cells