Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Damulin A | 1202868-74-3 | 96.9% | 783.00 g/mol | C42H70O13 | 10 MG
SDP

Catalog No. 5000298707
Encompass_Preferred

Damulin A is a triterpenoid saponin isolated from Gynostemma pentaphyllum with reported anticancer activity. It is supplied for research use only and is supported by analytical and safety documentation; consult the datasheet and SDS for handling, storage, and application details.

  • Purity 96.9% as listed on the product page.
  • Molecular weight 783.00 g/mol.
  • Chemical formula C42H70O13.
  • CAS number 1202868-74-3.
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.
  • Storage: sealed, away from moisture and light; in solvent store at -80°C for 6 months or -20°C for 1 month.
  • Analytical documents available including COA, HPLC, MS, NMR, and SDS.

Catalog No. 50-002-98707 Supplier Medchemexpress LLC Supplier No. HY1694110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Damulin A is a triterpenoid saponin isolated from Gynostemma pentaphyllum with reported anticancer activity. It is supplied for research use only and is supported by analytical and safety documentation; consult the datasheet and SDS for handling, storage, and application details.

  • Purity 96.9% as listed on the product page.
  • Molecular weight 783.00 g/mol.
  • Chemical formula C42H70O13.
  • CAS number 1202868-74-3.
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.
  • Storage: sealed, away from moisture and light; in solvent store at -80°C for 6 months or -20°C for 1 month.
  • Analytical documents available including COA, HPLC, MS, NMR, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.