Learn More
Cayman Chemical DAN Reagent 1ea
SDP
Catalog No. 502893819
Encompass_Preferred
An analytical reference standard that is structurally similar to known opioids intended for research and forensic applications
Catalog No. 50-289-3819 Supplier Cayman Chemical Supplier No. 7800701 EA
An analytical reference standard that is structurally similar to known opioids intended for research and forensic applications