Learn More
Medchemexpress LLC Danegaptide hydrochloride | 943133-81-1 | 99.49% | 327.76 | 1 ML
SDP
Catalog No. 5000346564
Encompass_Preferred
Danegaptide Hydrochloride (GAP-134 Hydrochloride) is a potent, selective, and orally active gap-junction modifier with an antiarrhythmic effect. It is for research use only and not sold to patients.
  • Potent, selective, and orally active gap-junction modifier.
  • Exhibits antiarrhythmic effect.
  • Evaluated for in vivo efficacy in the mouse CaCl2 model, significantly prolonging the time to conduction block.
  • Biologically active upon oral administration, reducing atrial fibrillation in a dog model without affecting heart rate, arterial blood pressure, or other electrocardiogram (ECG) parameters.
Catalog No. 50-003-46564 Supplier Medchemexpress LLC Supplier No. HY10913A10MM/1ML
Danegaptide Hydrochloride (GAP-134 Hydrochloride) is a potent, selective, and orally active gap-junction modifier with an antiarrhythmic effect. It is for research use only and not sold to patients.
  • Potent, selective, and orally active gap-junction modifier.
  • Exhibits antiarrhythmic effect.
  • Evaluated for in vivo efficacy in the mouse CaCl2 model, significantly prolonging the time to conduction block.
  • Biologically active upon oral administration, reducing atrial fibrillation in a dog model without affecting heart rate, arterial blood pressure, or other electrocardiogram (ECG) parameters.