Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Danger Barricade Tape

Catalog No. 19104305 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-305 Black on Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-305 Supplier Brady™ Supplier No. 91089
Add to Cart
Add to Cart

DANGER DO NOT ENTER

  • Use our Biodegradable Flagging Tape when hazardous marking is required in ecology conscious situations

Specifications

Color Black on Red
Width (English) 3 in.
Width (Metric) 127 mm
Length (English) 50 yd.
Packaging Quantity 150 ft./Roll
Certifications/Compliance OSHA 1910.144
Displayed Text DANGER DO NOT ENTER
Material 69% Wood Fiber
Type Barricade Tape
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.