Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Dangerous or Delicious: Using Chromatography to Examine Vaping
SDPSCI_ED

Catalog No. S52004 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S52004 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S52004 Supplier Edvotek™ Supplier No. 211

missing translation for 'validMainframeMsg'

In this experiment, students become medical researchers and investigate the chemical contents of three simulated e-liquids using thin layer chromatography.

  • Explore the health risks of e-cigarettes and vaping
  • Investigate the chemical contents of three simulated e-liquids
  • Perform Thin Layer Chromatography
  • Introduce chromatographic theory using two different solvent systems
  • Enough material for 8 separations
  • Time required: Spotting Plates - 20 minutes, TLC Separation - 5 minutes
  • Kit includes Instructions, samples, reagents and solvents, cellulose thin layer plates, 15mL tubes, & microcentrifuge tubes
  • All you need: 250 ml beakers, metric rulers, pencils, gloves & safety goggles
  • Some Components Require Refrigerator Storage

Specifications

Grade 9 to Undergraduate
Age 14 to 20
For Use With (Application) Perform Thin Layer Chromatography
Includes Instructions, samples, reagents and solvents, cellulose thin layer plates, 15 mL tubes, & microcentrifuge tubes
Requires 250 mL beakers, metric rulers, pencils, gloves & safety goggles
Content And Storage Some Components Require Refrigerator Storage
Product Type Chromatography Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.