Learn More
Medchemexpress LLC Danicopan (ACH-4471) | 1903768-17-1 | MFCD32669786 | 99.9% | 580.4 g/mol | C26H23BrFN7O3 | 50 MG
SDP
Catalog No. 5000227699
Encompass_Preferred
Danicopan (ACH-4471) is a selective, orally active small-molecule inhibitor of complement factor D, provided as a high-purity research compound for in vitro and preclinical studies. The material is supplied as a 50 mg solid with reported purity of approximately 99.9% and a molecular weight of 580.4 g/mol.
  • High purity suitable for research and analytical applications.
  • Selective factor D inhibition for complement pathway studies.
  • Supplied as a solid for convenient storage and formulation.
  • Well-characterized identity and molecular weight for reproducibility.
  • Appropriate for in vitro and preclinical pharmacology experiments.
Catalog No. 50-002-27699 Supplier Medchemexpress LLC Supplier No. HY11793050MG
Danicopan (ACH-4471) is a selective, orally active small-molecule inhibitor of complement factor D, provided as a high-purity research compound for in vitro and preclinical studies. The material is supplied as a 50 mg solid with reported purity of approximately 99.9% and a molecular weight of 580.4 g/mol.
  • High purity suitable for research and analytical applications.
  • Selective factor D inhibition for complement pathway studies.
  • Supplied as a solid for convenient storage and formulation.
  • Well-characterized identity and molecular weight for reproducibility.
  • Appropriate for in vitro and preclinical pharmacology experiments.