Learn More
MSC Danner Work Boot, Steel Toe, Nylon Upper, 1 ea
Catalog No. 5000795093
Encompass
This work boot is a military-style boot designed for men, featuring a steel toe for protection. It has a durable nylon upper and a rubber outsole, suitable for demanding environments.
  • Steel toe for impact and compression resistance
  • Nylon upper material for durability
  • Rubber outsole for traction
  • Scuff and abrasion-resistant properties
  • Fabric lining for comfort
  • Lace-up closure for a secure fit
  • Meets ASTM F2413-11 I/75 C/75 EH standards
  • Eight inch height
Catalog No. 50-007-95093 Supplier MSC Supplier No. 97845853
This work boot is a military-style boot designed for men, featuring a steel toe for protection. It has a durable nylon upper and a rubber outsole, suitable for demanding environments.
  • Steel toe for impact and compression resistance
  • Nylon upper material for durability
  • Rubber outsole for traction
  • Scuff and abrasion-resistant properties
  • Fabric lining for comfort
  • Lace-up closure for a secure fit
  • Meets ASTM F2413-11 I/75 C/75 EH standards
  • Eight inch height