Learn More
Medchemexpress LLC Danofloxacin-d3-1 | 1217683-55-0 | 360.40 | 5 MG
SDP
Catalog No. 5000452195
Encompass_Preferred
Danofloxacin-d3 is deuterium labeled Danofloxacin. It is an orally active quinolone antibiotic that targets bacterial DNA gyrase and inhibits bacterial DNA replication, transcription, and growth. It can be used for various bacterial infections caused by Escherichia coli, Mycoplasma, and other pathogens.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-52195 Supplier Medchemexpress LLC Supplier No. HYW011117S15MG
Danofloxacin-d3 is deuterium labeled Danofloxacin. It is an orally active quinolone antibiotic that targets bacterial DNA gyrase and inhibits bacterial DNA replication, transcription, and growth. It can be used for various bacterial infections caused by Escherichia coli, Mycoplasma, and other pathogens.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS