Learn More
Apexbio Technology LLC Danofloxacin Mesylate 119478-55-6 5g
SDP
Catalog No. 5000094365
Encompass_Preferred
Danofloxacin Mesylate (119478-55-6) is a small-molecule inhibitor targeting bacterial DNA gyrase and topoisomerase IV It is designed to inhibit these enzymes thereby impeding bacterial DNA replication and transcription Danofloxacin Mesylate exerts its biological activity primarily through inhibition of bacterial DNA gyrase and topoisomerase IV In microbiological assays Danofloxacin demonstrates antibacterial inhibition with a minimum inhibitory concentration (MIC90) of approximately 0 28 M Based on these pharmacological properties Danofloxacin Mesylate holds research potential in veterinary antimicrobial studies investigation of bacterial resistance mechanisms and comparative evaluation of antimicrobial activity
Catalog No. 50-000-94365 Supplier Apexbio Technology LLC Supplier No. B17205000
Danofloxacin Mesylate (119478-55-6) is a small-molecule inhibitor targeting bacterial DNA gyrase and topoisomerase IV It is designed to inhibit these enzymes thereby impeding bacterial DNA replication and transcription Danofloxacin Mesylate exerts its biological activity primarily through inhibition of bacterial DNA gyrase and topoisomerase IV In microbiological assays Danofloxacin demonstrates antibacterial inhibition with a minimum inhibitory concentration (MIC90) of approximately 0 28 M Based on these pharmacological properties Danofloxacin Mesylate holds research potential in veterinary antimicrobial studies investigation of bacterial resistance mechanisms and comparative evaluation of antimicrobial activity